accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSH3COOO

CAS RN 107802-80-2

IUPAC Name: 3,4,5-tris(benzyloxy)-2-methyl-6-(methylsulfanyl)oxane
Synonyms: 3,4,5-tris(benzyloxy)-2-methyl-6-(methylsulfanyl)oxane
Molecular Weight (g/mol): 464.62
Molecular Formula: C28H32O4S
InChi Key: DENOSPOKQXYGTJ-UHFFFAOYNA-N
SMILES: CSC1OC(C)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results