Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3,4-Tri-O-benzyl-1-thio-beta-L-fucopyranoside 95.0+%, TCI America™
SDP

Catalog No. M16281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16281G Supplier TCI America Supplier No. M16281G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent with >95.0% HPLC purity.

Chemical Identifiers

CAS 107802-80-2
Molecular Formula C28H32O4S
Molecular Weight (g/mol) 464.62
MDL Number MFCD06797141
InChI Key DENOSPOKQXYGTJ-NNVCNDCRSA-N
PubChem CID 11081093
IUPAC Name (2S,3R,4R,5S,6R)-2-methyl-6-methylsulfanyl-3,4,5-tris(phenylmethoxy)oxane
SMILES CC1C(C(C(C(O1)SC)OCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 1 g
Formula Weight 464.62
Percent Purity ≥95.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3,4-Tri-O-benzyl-1-thio-beta-L-fucopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.