accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONH3C

CAS RN 107869-45-4

IUPAC Name: 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
Synonyms: 10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
Molecular Weight (g/mol): 213.3
Molecular Formula: C10H15NO2S
InChi Key: ZAHOEBNYVSWBBW-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC11CS(=O)(=O)N=C1C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results