Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-10-Camphorsulfonimine 97.0+%, TCI America™
SDP

Catalog No. C13915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13915G Supplier TCI America Supplier No. C13915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107869-45-4
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.295
MDL Number MFCD00013315
InChI Key ZAHOEBNYVSWBBW-XCBNKYQSSA-N
Synonym (7R)-(+)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.0(1,5)]dec-4-ene 3,3-Dioxide, (3aR)-(+)-4,5,6,7-Tetrahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole 2,2-Dioxide
PubChem CID 98045278
SMILES CC1(C2CCC13CS(=O)(=O)N=C3C2)C

Specifications

Color White
Quantity 5 g
Formula Weight 213.30
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (+)-10-Camphorsulfonimine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.