CAS RN 108-10-1
Methyl iso-Butyl Ketone (Certified ACS), Fisher Chemical™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99%
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, ACS, 98.5+%
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99.5%, spectrophotometric grade
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, HPLC Grade, 99+%
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99+%, electronic use grade, residue free
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(C)=O
Methyl Isobutyl Ketone, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.
Methyl Isobutyl Ketone, Solstice
CAS: 108-10-1 PubChem CID: 7909 ChEBI: CHEBI:82344
Methyl Isobutyl Ketone ACS AR, Macron Fine Chemicals™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
Methyl Isobutyl Ketone ChromAR, Macron Fine Chemicals™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
Methyl-2-pentanone, 98.5% min., ACS, MilliporeSigma™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-Pentanone, OmniSolv™ MilliporeSigma™
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 98.5+%, ACS reagent
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
4-Methyl-2-pentanone, 99.5%, for analysis
CAS: 108-10-1 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 MDL Number: MFCD00008938 InChI Key: NTIZESTWPVYFNL-UHFFFAOYSA-N Synonym: 4-methyl-2-pentanone,methyl isobutyl ketone,isopropylacetone,isobutyl methyl ketone,hexone,mibk,2-pentanone, 4-methyl,2-methyl-4-pentanone,4-methyl-2-oxopentane,methylisobutylketon PubChem CID: 7909 ChEBI: CHEBI:82344 IUPAC Name: 4-methylpentan-2-one SMILES: CC(C)CC(=O)C
- 1
- 2