accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2COCH3O

CAS RN 108-22-5

IUPAC Name: prop-1-en-2-yl acetate
Synonyms: 1-propen-2-ol acetate
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: HETCEOQFVDFGSY-UHFFFAOYSA-N
SMILES: CC(=C)OC(C)=O
Boiling Point 
  • (1)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)