Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropenyl acetate

Catalog No. 117781250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-778-1250ML 250 mL
11-778-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-778-1250ML Supplier Sigma Aldrich Supplier No. 117781250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isopropenyl acetate forms copolyperoxides of different compositions with indene by the free radical-initiated oxidative copolymerization.

Specifications

CAS 108-22-5
Density 0.922 g/mL at 20°C, 0.909 g/mL (at 25°C (literature))
Boiling Point 94°C (lit.)
Molecular Formula C5H8O2
Refractive Index n20/D 1.401 (literature)
Linear Formula CH3CO2C(CH3)=CH2
MDL Number MFCD00008709
Quantity 250 mL
Synonym 1-Methylvinyl acetate
Molecular Weight (g/mol) 100.12
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.