accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-29-2

IUPAC Name: 5-methyloxolan-2-one
Synonyms: gamma-valerolactone 4-valerolactone gamma-valeryllactone gamma-valerolakton dihydr-5-methyl-2(3H)-furanone gamma-pentanolactone 4-methyl-gamma-butyrolactone gamma-pentalactone 4-pentanolide 4-hydroxyvaleric acid lactone
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: GAEKPEKOJKCEMS-UHFFFAOYNA-N
SMILES: CC1CCC(=O)O1
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)