Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich gamma-Valerolactone

Catalog No. V403500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V403500G 500 g
V403100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V403500G Supplier Sigma Aldrich Supplier No. V403500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

gamma-Valerolactone

Specifications

CAS 108-29-2
Melting Point -31°C (lit.)
Density 1.05 g/mL (at 25°C (literature))
Boiling Point 207°C to 208°C (lit.), 82°C to 85°C (10 mmHg)
Molecular Formula C5H8O2
Linear Formula C5H8O2
Refractive Index n20/D 1.432 (literature)
MDL Number MFCD00005400
Quantity 500 g
Synonym gamma-Valerolactone; gamma-Valerolactone; gamma-Methyl-gamma-butyrolactone; gamma-Methyl-gamma-butyrolactone; 4,5-Dihydro-5-methyl-2(3 H)-furanone; 4-Hydroxypentanoic acid lactone
Molecular Weight (g/mol) 100.12
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.