CAS RN 108-32-7
HYDRANAL™ - Water Standard 0.1 PC, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Solstice
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, 99.5%
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, 99%
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, MP Biomedicals™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene Carbonate, BAKER™, J.T. Baker™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene Carbonate 98.0+%, TCI America™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Sigma Aldrich Propylene carbonate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), Solstice
Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: 1,2-Propanediol cyclic carbonate,4-Methyl-1,3-dioxolan-2-one PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C
Propylene carbonate, CHROMASOLV™, for HPLC, 99.7%, Solstice
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1