Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene carbonate, MP Biomedicals™

Catalog No. ICN15641594 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15641594 5 kg
ICN15641590 500 g
ICN15641591 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15641594 Supplier MP Biomedicals, Inc Supplier No. 0215641594
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Color Colorless
Density ∽1.19g/mL
Quantity 5 kg
Formula Weight 102.09
Physical Form Liquid
Chemical Name or Material Propylene carbonate, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.