accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-37-2

IUPAC Name: 1-bromo-3-chlorobenzene
Synonyms: M-bromochlorobenzene
Molecular Weight (g/mol): 191.45
Molecular Formula: C6H4BrCl
InChi Key: JRGGUPZKKTVKOV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Br)=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)