accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2N

CAS RN 108-44-1

IUPAC Name: 3-methylaniline
Synonyms: M-toluidine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: JJYPMNFTHPTTDI-UHFFFAOYSA-N
SMILES: CC1=CC=CC(N)=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (8)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)