Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich m-Toluidine

Catalog No. 5112185ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-121-85ML 5 mL
51-121-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 51-121-85ML Supplier Sigma Aldrich Supplier No. 5112185ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • M-toluidine (mt) is an aniline derivative that behaves as a fragile glass forming liquid
  • Its dielectric relaxation process, glass transition relaxation, relaxation modes and slow secondary relaxation have been studied
  • The analysis of mt crystalline phase by powder x-ray diffraction shows that it belongs to monoclinic space group, p21/c
  • Its structure is held together by n-h...n hydrogen bonds and van der waals interactions.m-toluidine may be used to form pyrrole/m-toluidine copolymers via oxidative polymerization

Specifications

CAS 108-44-1
Density 0.999 g/mL (at 25°C (literature))
Boiling Point 203°C to 204°C (lit.)
Molecular Formula C7H9N
Refractive Index n20/D 1.567 (literature)
Linear Formula CH3C6H4NH2
MDL Number MFCD00007808
Quantity 5 mL
Synonym 3-Methylaniline
Molecular Weight (g/mol) 107.15
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.