accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOCH3Cl

CAS RN 108-60-1

IUPAC Name: 1-chloro-2-[(1-chloropropan-2-yl)oxy]propane
Synonyms: 1-chloro-2-[(1-chloropropan-2-yl)oxy]propane
Molecular Weight (g/mol): 171.06
Molecular Formula: C6H12Cl2O
InChi Key: QCFYJCYNJLBDRT-UHFFFAOYNA-N
SMILES: CC(CCl)OC(C)CCl
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Element 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results