Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Benzoate Solution, 5000μg/mL in hexane, Ultra Scientific

Catalog No. USIST4001
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-4001 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-4001 Supplier Agilent Technologies Supplier No. IST4001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-60-1 , 110-54-3
Molecular Formula C6H12Cl2O
Molecular Weight (g/mol) 171.061
InChI Key QCFYJCYNJLBDRT-UHFFFAOYSA-N
PubChem CID 7944
ChEBI CHEBI:82409
IUPAC Name 1-chloro-2-(1-chloropropan-2-yloxy)propane
SMILES CC(CCl)OC(C)CCl

Specifications

CAS Max % 99.24, 0.76
Analyte or Component Names n-hexane,benzyl benzoate
Element n-hexane, Benzyl Benzoate
Solvent or Matrix Hexane
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Benzyl Benzoate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.