CAS RN 108-65-6
Propylene Glycol Methyl Ether Acetate, Spectrum™ Chemical
CAS: 108-65-6 Molecular Weight (g/mol): 132.16
2-(1-Methoxy)propyl acetate, 99%
CAS: 108-65-6 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.16 MDL Number: MFCD00038500 InChI Key: LLHKCFNBLRBOGN-UHFFFAOYSA-N Synonym: 1-methoxy-2-propyl acetate,pgmea,2-acetoxy-1-methoxypropane,propylene glycol monomethyl ether acetate,propylene glycol methyl ether acetate,1-methoxy-2-acetoxypropane,2-propanol, 1-methoxy-, acetate,2-methoxy-1-methylethyl acetate,propyleneglycol monomethyl ether acetate,acetic acid, 2-methoxy-1-methylethyl ester PubChem CID: 7946 IUPAC Name: 1-methoxypropan-2-yl acetate SMILES: CC(COC)OC(=O)C
1-Methoxy-2-propanol Acetate, 97%, Spectrum™ Chemical
CAS: 108-65-6 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.16 InChI Key: LLHKCFNBLRBOGN-UHFFFAOYNA-N IUPAC Name: 1-methoxypropan-2-yl acetate SMILES: COCC(C)OC(C)=O
1,2-Propanediol monomethyl ether acetate, 99%, stab. with 50ppm BHT
CAS: 108-65-6 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.159 MDL Number: MFCD00038500 InChI Key: LLHKCFNBLRBOGN-UHFFFAOYSA-N Synonym: 1-methoxy-2-propyl acetate,pgmea,2-acetoxy-1-methoxypropane,propylene glycol monomethyl ether acetate,propylene glycol methyl ether acetate,1-methoxy-2-acetoxypropane,2-propanol, 1-methoxy-, acetate,2-methoxy-1-methylethyl acetate,propyleneglycol monomethyl ether acetate,acetic acid, 2-methoxy-1-methylethyl ester PubChem CID: 7946 IUPAC Name: 1-methoxypropan-2-yl acetate SMILES: CC(COC)OC(=O)C
Propylene Glycol 1-Monomethyl Ether 2-Acetate 98.0+%, TCI America™
CAS: 108-65-6 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.159 MDL Number: MFCD00038500 InChI Key: LLHKCFNBLRBOGN-UHFFFAOYSA-N Synonym: 1-methoxy-2-propyl acetate,pgmea,2-acetoxy-1-methoxypropane,propylene glycol monomethyl ether acetate,propylene glycol methyl ether acetate,1-methoxy-2-acetoxypropane,2-propanol, 1-methoxy-, acetate,2-methoxy-1-methylethyl acetate,propyleneglycol monomethyl ether acetate,acetic acid, 2-methoxy-1-methylethyl ester PubChem CID: 7946 IUPAC Name: 1-methoxypropan-2-yl acetate SMILES: CC(COC)OC(=O)C
Sigma Aldrich Propylene Glycol Monomethyl Ether Acetate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Propylene Glycol Methyl Ether Acetate, SPEX CertiPrep™
CAS: 108-65-6 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.159 InChI Key: LLHKCFNBLRBOGN-UHFFFAOYSA-N PubChem CID: 7946 IUPAC Name: 1-methoxypropan-2-yl acetate SMILES: CC(COC)OC(=O)C