accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108166-03-6

IUPAC Name: 2-methylquinoline-6-carbaldehyde
Synonyms: 2-methylquinoline-6-carbaldehyde
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: WAPQHBBEWPVBTO-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(C=O)C=CC2=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results