Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylquinoline-6-carboxaldehyde, 95%, Thermo Scientific™

Catalog No. AAH63651MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63651MD 250 mg
AAH6365103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63651MD Supplier Thermo Scientific Chemicals Supplier No. H63651MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108166-03-6
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.20
MDL Number MFCD08060502
InChI Key WAPQHBBEWPVBTO-UHFFFAOYSA-N
Synonym 2-methyl-6-quinolinecarbaldehyde, 2-methyl-6-quinolinecarboxaldehyde, 2-methylquinoline-6-carboxaldehyde, 6-quinolinecarboxaldehyde,2-methyl, 6-quinolinecarboxaldehyde, 2-methyl, acmc-1bu0b, 6-formyl-2-methylquinoline, 2-methyl-quinoline-6-carbaldehyde
PubChem CID 7537486
IUPAC Name 2-methylquinoline-6-carbaldehyde
SMILES CC1=CC=C2C=C(C=O)C=CC2=N1

Specifications

Melting Point 96°C to 99°C
Color Yellow
Quantity 250 mg
UN Number UN2811
Formula Weight 171.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Methylquinoline-6-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.