accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOPOHOOPOOHHOOHOHOPOOHHONa

CAS RN 108321-05-7

IUPAC Name: {[({3,4-dihydroxy-5-[(phosphonooxy)methyl]oxolan-2-yl}oxy)(hydroxy)phosphoryl]oxy}phosphonic acid sodium
Synonyms: ({3,4-dihydroxy-5-[(phosphonooxy)methyl]oxolan-2-yl}oxy(hydroxy)phosphoryl)oxyphosphonic acid sodium
Molecular Weight (g/mol): 413.06
Molecular Formula: C5H13NaO14P3
InChi Key: CFIMWVBJOCHTCA-UHFFFAOYNA-N
SMILES: [Na].OC1C(O)C(OP(O)(=O)OP(O)(O)=O)OC1COP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results