accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108374-32-9

IUPAC Name: platinum(2+) tetraamine dichloride
Synonyms: platinum(2+) tetraamine dichloride
Molecular Weight (g/mol): 334.11
Molecular Formula: Cl2H12N4Pt
InChi Key: KHCPSOMSJYAQSY-UHFFFAOYSA-L
SMILES: N.N.N.N.[Cl-].[Cl-].[Pt++]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)