Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraammineplatinum(II) Chloride Hydrate

Catalog No. 2759055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-590-55G 5 g
27-590-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 27-590-55G Supplier Sigma Aldrich Supplier No. 2759055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetraammineplatinum(II) chloride hydrate can be used as a source of platinum:To deposit layer of Pt electrode on Nafion membranes.To be used as a coupling catalyst in the electron beam technology for the decomposition of volatile organic compounds.

Specifications

CAS 108374-32-9
Molecular Formula H12Cl2N4Pt -+ xH2O
Linear Formula Pt(NH3)4Cl2-+xH2O
MDL Number MFCD00149947
Quantity 5 g
Molecular Weight (g/mol) 334.11 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.