accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNNH2

CAS RN 108468-00-4

IUPAC Name: tert-butyl N-{[4-(aminomethyl)phenyl]methyl}carbamate
Synonyms: tert-butyl N-{[4-(aminomethyl)phenyl]methyl}carbamate
Molecular Weight (g/mol): 236.32
Molecular Formula: C13H20N2O2
InChi Key: NUANLVJLUYWSER-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(CN)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)