accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COHCH3OH3COHH2OZn(E/Z)(E/Z)

CAS RN 108503-47-5

IUPAC Name: bis(4-hydroxypent-3-en-2-one) hydrate zinc
Synonyms: bis(4-hydroxypent-3-en-2-one) hydrate zinc
Molecular Weight (g/mol): 283.63
Molecular Formula: C10H18O5Zn
InChi Key: JZOZHSRJFJNUOU-UHFFFAOYSA-N
SMILES: O.[Zn].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)