accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1085181-82-3

IUPAC Name: 2-bromo-5-iodo-3-octylthiophene
Synonyms: 2-bromo-5-iodo-3-octylthiophene
Molecular Weight (g/mol): 401.14
Molecular Formula: C12H18BrIS
InChi Key: WYOLKTBMZYMZBF-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=C(Br)SC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results