Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-iodo-3-n-octylthiophene (stabilized with Copper chip) 96.0+%, TCI America™
SDP

Catalog No. B45841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45841G Supplier TCI America Supplier No. B45841G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1085181-82-3
Molecular Formula C12H18BrIS
Molecular Weight (g/mol) 401.144
InChI Key WYOLKTBMZYMZBF-UHFFFAOYSA-N
PubChem CID 91972177
IUPAC Name 2-bromo-5-iodo-3-octylthiophene
SMILES CCCCCCCCC1=C(SC(=C1)I)Br

Specifications

Color Yellow
Boiling Point 152°C
Quantity 1 g
Formula Weight 401.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-iodo-3-n-octylthiophene (stabilized with Copper chip)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.