accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1086-80-2

IUPAC Name: 7,8-dimethyl-1H,2H,3H,4H-benzo[g]pteridine-2,4-dione
Synonyms: 7,8-dimethylalloxazine 7,8-dimethyl- (8CI) alloxazine riboflavin lumichrome lumichrome 6,7-dimethylalloxazine
Molecular Weight (g/mol): 242.24
Molecular Formula: C12H10N4O2
InChi Key: ZJTJUVIJVLLGSP-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2N=C3C(=O)NC(=O)NC3=NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results