Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7,8-Dimethylalloxazine 98.0+%, TCI America™
SDP

Catalog No. D1066100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1066100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1066100MG Supplier TCI America Supplier No. D1066100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1086-80-2
Molecular Formula C12H10N4O2
Molecular Weight (g/mol) 242.24
MDL Number MFCD00005021
InChI Key ZJTJUVIJVLLGSP-UHFFFAOYSA-N
Synonym Lumichrome
PubChem CID 5326566
ChEBI CHEBI:37323
IUPAC Name 7,8-dimethyl-1H,2H,3H,4H-benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N=C3C(=O)NC(=O)NC3=NC2=C1

Specifications

Quantity 100 mg
Formula Weight 242.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7,8-Dimethylalloxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.