accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONFBr

CAS RN 1086600-40-9

IUPAC Name: 4-[(2-bromo-4-fluorophenyl)methyl]morpholine
Synonyms: 4-[(2-bromo-4-fluorophenyl)methyl]morpholine
Molecular Weight (g/mol): 274.13
Molecular Formula: C11H13BrFNO
InChi Key: JRIHSKPSHQDJMT-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(CN2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results