CAS RN 1087-97-4
CAS RN 1087-97-4
IUPAC Name:
1,3-diethyl 2-benzoylpropanedioate
Synonyms:
1,3-diethyl 2-benzoylpropanedioate
Molecular Weight (g/mol):
264.28
Molecular Formula:
C14H16O5
InChi Key:
RIQBATDJIKIMBM-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(C(=O)OCC)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Diethyl Benzoylmalonate 97.0+%, TCI America™
CAS: 1087-97-4 Molecular Formula: C14H16O5 Molecular Weight (g/mol): 264.277 MDL Number: MFCD00059381 InChI Key: RIQBATDJIKIMBM-UHFFFAOYSA-N Synonym: Benzoylmalonic Acid Diethyl Ester PubChem CID: 569310 IUPAC Name: diethyl 2-benzoylpropanedioate SMILES: CCOC(=O)C(C(=O)C1=CC=CC=C1)C(=O)OCC