Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Benzoylmalonate 97.0+%, TCI America™
SDP

Catalog No. B020925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B020925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B020925G Supplier TCI America Supplier No. B020925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1087-97-4
Molecular Formula C14H16O5
Molecular Weight (g/mol) 264.277
MDL Number MFCD00059381
InChI Key RIQBATDJIKIMBM-UHFFFAOYSA-N
Synonym Benzoylmalonic Acid Diethyl Ester
PubChem CID 569310
IUPAC Name diethyl 2-benzoylpropanedioate
SMILES CCOC(=O)C(C(=O)C1=CC=CC=C1)C(=O)OCC

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 g
Formula Weight 264.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Benzoylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.