accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNHNHCl

CAS RN 1087737-96-9

IUPAC Name: 5,6-dichloro-1H-1,3-benzodiazole hydrochloride
Synonyms: 5,6-dichloro-1H-1,3-benzodiazole hydrochloride
Molecular Weight (g/mol): 223.48
Molecular Formula: C7H5Cl3N2
InChi Key: CKILENXYURTTFP-UHFFFAOYSA-N
SMILES: Cl.ClC1=C(Cl)C=C2N=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)