Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichlorobenzimidazole Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D42955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42955G 5 g
D42951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42955G Supplier TCI America Supplier No. D42955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1087737-96-9
Molecular Formula C7H5Cl3N2
Molecular Weight (g/mol) 223.481
MDL Number MFCD16810292
InChI Key CKILENXYURTTFP-UHFFFAOYSA-N
PubChem CID 24187360
IUPAC Name 5,6-dichloro-1H-benzimidazole;hydrochloride
SMILES C1=C2C(=CC(=C1Cl)Cl)N=CN2.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 223.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichlorobenzimidazole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.