accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHN

CAS RN 108963-20-8

IUPAC Name: N,1-dibenzylpyrrolidin-3-amine
Synonyms: N,1-dibenzylpyrrolidin-3-amine
Molecular Weight (g/mol): 266.39
Molecular Formula: C18H22N2
InChi Key: BHEXKVGLZMEJRQ-UHFFFAOYNA-N
SMILES: C(NC1CCN(CC2=CC=CC=C2)C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results