CAS RN 109-46-6
IUPAC Name:
1,3-dibutylthiourea
Synonyms:
N,N'-di-n-butylthiourea
Molecular Weight (g/mol):
188.33
Molecular Formula:
C9H20N2S
InChi Key:
KFFQABQEJATQAT-UHFFFAOYSA-N
SMILES:
CCCCNC(=S)NCCCC