accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109-56-8

IUPAC Name: 2-[(propan-2-yl)amino]ethan-1-ol
Synonyms: 2-(isopropylamino)ethanol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: RILLZYSZSDGYGV-UHFFFAOYSA-N
SMILES: CC(C)NCCO
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)