Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylaminoethanol, MP Biomedicals

Catalog No. ICN208747 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208747 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208747 Supplier MP Biomedicals, Inc Supplier No. 0520874710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109-56-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
InChI Key RILLZYSZSDGYGV-UHFFFAOYSA-N
Synonym 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine
PubChem CID 7994
IUPAC Name 2-(propan-2-ylamino)ethanol
SMILES CC(C)NCCO

Specifications

Quantity 10 g
Formula Weight 103.7
Grade Not Available
Chemical Name or Material 2-Isopropylaminoethanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.