accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2NN

CAS RN 109-82-0

IUPAC Name: 2-(methylideneamino)acetonitrile
Synonyms: 2-(methylideneamino)acetonitrile
Molecular Weight (g/mol): 68.08
Molecular Formula: C3H4N2
InChi Key: GFZMFCVDDFHSJK-UHFFFAOYSA-N
SMILES: C=NCC#N
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)