Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyleneiminoacetonitrile, 98%, Thermo Scientific™

Catalog No. AAA1716222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1716222 100 g
AAA1716214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1716222 Supplier Thermo Scientific Chemicals Supplier No. A1716222

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-82-0
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00001889
InChI Key GFZMFCVDDFHSJK-UHFFFAOYSA-N
Synonym methyleneaminoacetonitrile, methylenaminoacetonitrile, maan, acetonitrile, methyleneamino, methyleniminoacetonitrile, glycinonitrile, n-methylene, n-methyleneglycinonitrile, 2-methylideneamino acetonitrile, methylenamino acetonitrile, usaf do-5
PubChem CID 8015
IUPAC Name 2-(methylideneamino)acetonitrile
SMILES C=NCC#N

Specifications

Melting Point 128°C to 129°C
Linear Formula CH2=NCH2CN
Quantity 100 g
Beilstein 1848155
Merck Index 14,6057
Formula Weight 68.08
Percent Purity 98%
Chemical Name or Material Methyleneiminoacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-709-5
RTECSNumber MC1925000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.