accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHHO

CAS RN 109-83-1

IUPAC Name: 2-(methylamino)ethan-1-ol
Synonyms: methylethanolamine N-methyl-2-ethanolamine methylethylolamine monomethyl-aminoaethanol N-methyl-2-aminoethanol
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: OPKOKAMJFNKNAS-UHFFFAOYSA-N
SMILES: CNCCO
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)