Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylamino)ethanol, 99%

Catalog No. AC158260025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158260025 2.5 kg Glass bottle
AC158265000 500 g Glass bottle
AC158260100 10 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158260025 Supplier Thermo Scientific Chemicals Supplier No. 158260025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 109-83-1 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-83-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00002839
InChI Key OPKOKAMJFNKNAS-UHFFFAOYSA-N
Synonym 2-methylamino ethanol, n-methylethanolamine, methylethanolamine, ethanol, 2-methylamino, methylethylolamine, n-methylaminoethanol, n-methylmonoethanolamine, n-monomethylethanolamine, 2-hydroxyethyl methylamine, monomethylaminoethanol
PubChem CID 8016
ChEBI CHEBI:21763
IUPAC Name 2-(methylamino)ethanol
SMILES CNCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -3.0°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 159.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Linear Formula CH3NHCH2CH2OH
Quantity 2.5 kg
Beilstein 04, 276
Specific Gravity 0.93
Merck Index 15, 6089
Solubility Information Solubility in water: miscible
Viscosity 13 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Methylamino)ethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-710-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.