accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHHO

CAS RN 109-84-2

IUPAC Name: 2-hydrazinylethan-1-ol
Synonyms: 2-hydroxyethylhydrazine epa pesticide chemical code 055502 brombloom caswell no. 488A BOH HEH ethanolhydrazine omaflora
Molecular Weight (g/mol): 76.1
Molecular Formula: C2H8N2O
InChi Key: GBHCABUWWQUMAJ-UHFFFAOYSA-N
SMILES: NNCCO
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (1)
Boiling Point 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)