Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydrazinoethanol 80.0+%, TCI America™
SDP

Catalog No. H106725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H106725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H106725G Supplier TCI America Supplier No. H106725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109-84-2
Molecular Formula C2H8N2O
Molecular Weight (g/mol) 76.099
MDL Number MFCD00007623
InChI Key GBHCABUWWQUMAJ-UHFFFAOYSA-N
Synonym 2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
PubChem CID 8017
ChEBI CHEBI:50098
IUPAC Name 2-hydrazinylethanol
SMILES C(CO)NN

Specifications

Color Colorless
Quantity 25 g
UN Number 2810
Formula Weight 76.10
Percent Purity ≥80.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Hydrazinoethanol
EINECSNumber (9)-1892
RTECSNumber KL2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.