accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109-85-3

IUPAC Name: 2-methoxyethan-1-amine
Synonyms: ethanamine, 2-methoxy-
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: ASUDFOJKTJLAIK-UHFFFAOYSA-N
SMILES: COCCN
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)