accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109-88-6

MgCH3OCH3O

CAS RN 109-88-6

IUPAC Name: magnesium(2+) dimethanolate
Synonyms: magnesium(2+) bis(methoxide)
Molecular Weight (g/mol): 86.37
Molecular Formula: C2H6MgO2
InChi Key: CRGZYKWWYNQGEC-UHFFFAOYSA-N
SMILES: [Mg++].C[O-].C[O-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)