Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Magnesium methoxide solution

Catalog No. 335657500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-565-7500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-565-7500ML Supplier Sigma Aldrich Supplier No. 335657500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed in a sol-gel preparation of nanocrystalline MgO with 1.8-35 nm particle sizes.

Specifications

CAS 109-88-6
Density 0.816 g/mL (at 25°C)
Molecular Formula C2H6MgO2
Refractive Index n20/D 1.3381
Linear Formula Mg(OCH3)2
Quantity 500 mL
MDL Number MFCD00053671
Molecular Weight (g/mol) 86.37
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.