accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNH3C

CAS RN 109-89-7

IUPAC Name: diethylamine
Synonyms: diethylamine
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: HPNMFZURTQLUMO-UHFFFAOYSA-N
SMILES: CCNCC
Molecular Weight (g/mol) 
  • (19)
  • (10)
Quantity 
  • (5)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (10)
  • (9)
  • (5)
Physical Form 
  • (16)
Grade 
  • (1)
  • (2)
  • (7)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (5)
  • (1)
  • (10)
  • (5)
  • (1)
  • (12)
  • (19)
  • (10)
  • (5)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (10)
  • (9)
  • (5)
  • (16)
  • (1)
  • (2)
  • (7)
  • (5)
  • (1)
  • (5)
  • (4)
  • (5)
  • (1)