Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylamine, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18602198 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-198 200 L Amber Glass Bottle
18-602-194 500 mL Amber Glass Bottle
18-602-197 2.5 L Amber Glass Bottle
18-602-195 4 L Amber Glass Bottle
18-602-196 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-198 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DI117200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DI117, 109-89-7, C4H11N

Spectrum™ Chemical Diethylamine, Reagent is a secondary amine that is a flammable, volatile, strongly alkaline, colorless liquid that has a strong unpleasant smell. It is used in the production of rubber, resin, dyes and pharmaceuticals and as a corrosion inhibiter. Its Reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
IUPAC Name diethylamine
SMILES CCNCC

Specifications

CAS Max % 100%
Boiling Point 55.5°C/132°F
Packaging Amber Glass Bottle
Quantity 200 L
UN Number UN1154
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Diethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.