accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109-98-8

IUPAC Name: 4,5-dihydro-1H-pyrazole
Synonyms: pyrazoline
Molecular Weight (g/mol): 70.1
Molecular Formula: C3H6N2
InChi Key: MCGBIXXDQFWVDW-UHFFFAOYSA-N
SMILES: C1CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results