Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyrazoline 95.0+%, TCI America™
SDP

Catalog No. P05475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P05475G Supplier TCI America Supplier No. P05475G

Specifications

CAS 109-98-8
Color Colorless
Boiling Point 144°C
Molecular Formula C3H6N2
Quantity 5 g
MDL Number MFCD00059715
UN Number 1993
InChI Key MCGBIXXDQFWVDW-UHFFFAOYSA-N
SMILES C1CNN=C1
IUPAC Name 4,5-dihydro-1H-pyrazole
Molecular Weight (g/mol) 70.095
PubChem CID 66962
Formula Weight 70.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pyrazoline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.